Would you like to log in again?
Biox Cell Scrapers and Lifters
Supplier: TradeWinds (TWD)
Biox
BX70-2180 BX70-1250 BX70-1180
75799-934CS 73.4 USD InStock
75799-934 75799-936 75799-938
Biox Cell Scrapers and Lifters
Cell Scrapers
Cell Scrapers feature a uniquely designed handle and blade which reduces cell damage during the harvesting process.
  • Non-pyrogenic
  • Polyethylene construction
  • Pivoting blade makes precise cutting and scraping movements
  • Disposable
  • Pre-sterilized and individually wrapped
Our scrapers have pivot blades for precise cutting and scraping and our lifters are one sturdy piece for improved control.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty