Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cell Strainers

Supplier: TradeWinds (TWD)
BX15-1070 BX15-1100 BX15-1040
75799-944CS 224.78 USD InStock
75799-944 75799-940 75799-942
Cell Strainers
Cell Strainers
Cell Strainers feature a strong nylon mesh for optimal performance in a variety of applications.

  • Polypropyelene frame with nylon mesh
  • Gamma radiation sterilized
  • Color coded for easy pore size identification
  • Individually wrapped
  • Applications in cell culture and filtration of various cells

Perfect fit for 50 ml centrifuge tubes and color coded according to the pore size.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty