Would you like to log in again?
Biox Cell Spreaders
Supplier: TradeWinds (TWD)
Biox
BX65-1001 BX65-1010
75799-960CS 125.92 USD InStock
75799-960 75799-958
Biox Cell Spreaders
Spreaders Cell Spreaders
Cell Spreaders are ideal for the even spreading of bacterial cultures.
  • Polystyrene construction
  • L-Shape design
  • Disposable
  • Smooth spreading surface with slight upward angle reduces the risk of damaging agar surfaces
  • Gamma radiation sterilized
  • Individually wrapped
L-Shaped cell spreaders allows for application on the entire culture dish or plate surface.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty