Would you like to log in again?
PCR Tubes
Supplier: TradeWinds (TWD)
60-0082 60-0002 BX60-0008
75799-966CS 481.43 USD InStock
75799-966 75799-968 75799-972
PCR Tubes
Tubes PCR Tubes
PCR tubes are ideally suited for real-time PCR applications.
  • Prime virgin PP construction
  • Temperature: stable from –20 to 100 °C (–4 to 212 °F)
  • Tubes withstand centrifugation of 6000 RCF
  • Autoclavable in open state
  • Ultra-thin wall tubes
Rnase-, Dnase-, and endotoxin free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty