Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethyleneimine, M.W. Mn ~ 10,000, branched

Supplier: POLYSCIENCES INC-RESEARCH CHEM
Synonyms: PolyethyleniminePEICorcatPolymin PEthyleneimine PolymerAziridinePoly(ethyleneimine)

19850-500 19850-25 19850-100
75800-204EA 154.45 USD InStock
75800-204 75800-206 75800-172
Polyethyleneimine, M.W. Mn ~ 10,000, branched
Polyethyleneimine
Molecular Weight: 10,000 (bPEI 10000)
Density: 1.029-1.038
Viscosity: 40,000 - 150,000 cps
Soluble In: water, lower alcohols, glycols, and THF
Resin Content (wt%): 99%
Appearance: clear light yellow viscous liquid

  • Highly branched liquid water soluble polyamine
  • Contains primary, secondary, and tertiary amine groups in approximately 25/50/25 ratio
  • Auxillary agent intensifying manufacturing prcesses

It is widely used in paper-making, water treatment, detergents, adhesives, and cosmetics.
Branched Polyethylenimine, Mw 10,000 (bPEI 10000) is highly branched liquid water soluble polyamine with high cationic charge density. bPEI 10000 contains primary, secondary, and tertiary amine groups in approximately 25/50/25 ratio. It acts as an auxiliary agent intensifying manufacturing processes and improving quality of the final products. Extensively investigated as non-viral vector carriers.


Formula: (CH₂CH₂NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty