Would you like to log in again?
VWR® TPX Measuring Cylinders, Class B
Supplier: VWR
VWR®
346080 346020 346030 346060 346010 346070 346050 346040
75800-224CS 245.13 USD InStock
75800-224 75800-226 75800-228 75800-218 75800-220 75800-230 75800-222 75800-232
VWR® TPX Measuring Cylinders, Class B
Cylinders Measuring Cylinders
These VWR® Measuring Cylinders are manufactured from translucent polypropylene.
  • The cylinders feature a stable hexagonal base and a pouring spout
  • All cylinders also feature molded graduations for improved accuracy and consistency
When used continuously, the cylinders have a working temperature range of 0 to 121 °C (32 to 248 °F).
Certifications: Products are Class "B" as per DIN 12681 / ISO 6706. Resin meets US FDA 21 CFR requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty