Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Branson SFX150 Sonifier® Cell Disruptor with Handheld Converter and 1/8" Microtip, 230V

Supplier: Branson

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

101-063-1097R
75800-282EA 5892.74 USD InStock
75800-282
Branson SFX150 Sonifier® Cell Disruptor with Handheld Converter and 1/8" Microtip, 230V
Homogenizers Ultrasonic Cell Disruptors
The SFX150 is designed to bring the industry's most advanced sample-processing capabilities to your laboratory

Branson's handheld converter is designed with the user in mind with an ergonomic grip for comfort and control, and a recessed pushbutton and LED indicator for assured operation.

True temperature control allows the user to specify a temperature range for the sample, the Sonifier will automatically adjust the ultrasonic pulse length to prevent the sample from overheating (requires optional temperature probe).

Advanced control modes includes time and energy mode, energy mode measured in joules, either continuously or in individual pulses. Process monitoring on a scrollable, digital screen provides updates of key variables including power level, energy usage, sample temperature, and experiment progress. For assured repeatability and precision on a greater scale, user can store up to 20 sample-processing programs. Program parameters include continuous or pulsed ultrasonics; time, energy, or temperature control modes; pulse on-time/energy, off-time and total on-time/energy; or amplitude as a percentage. Microtip limit 70%. Other accessories also available.

Applications include sample preparation, cell disruption, nanoparticle disperson, mixing, homogenization, DNA, cell lysis, protein extration.

Geared toward low-volume samples ranging from 0.2 to 150mL, the Sonifier SFX150 delivers up to 150 watts of power. And at 40kHz, it's quiet enough to operate without an enclosure. Unit comes with a 1/8" microtip, 240v, CE.

Certifications: CSA, UL, CE ISO9001, ISO14001.

Ordering information: 3 year warranty. Includes 40kHz Handheld converter, 1/8" microtip, power cord, manual, wrench set.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty