Would you like to log in again?
Nunc™ Edge 2.0 96-Well Plates, Thermo Scientific
Supplier: Thermo Scientific
167425 167554 267576 267544 267556 267427 267566 167542 267578 167574
75800-400CS 83.18 USD InStock
75800-400 75800-388 75800-402 75800-390 75800-392 75800-418 75800-394 75800-396 75800-398 75800-420
Nunc™ Edge 2.0 96-Well Plates, Thermo Scientific
Microplates Cell Culture Plates
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate.
  • Minimizes risks
  • Enhances productivity
  • Lowers cost
  • Reduces waste
When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays
The Edge 2.0 Plate provides an ideal, efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect. Uniquely designed with a surrounding moat that serves as an evaporation barrier, enabling researchers to expand their microplate cultures to all 96 wells without concerns of evaporation. Following incubation periods of up to four days, Edge 2.0 yields consistent, viable cells across the entire 96-well plate, leading to improved productivity for cell- based assays. Enabling the use of all 96 wells permits experiment efficiencies that can result in potentially less cost and related product waste.
Nunclon™ Delta cell culture surface promotes cell attachment and growth; untreated surface options available. Filling perimeter reservoir with 1.7 sterile water or media helps o ensure well-to-well consistency and cell viability. Overall plate evaporation rate reduced to <2% after four days of incubation. Total well volume = 400 µL. Using the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader, the filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Certifications: All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty