Would you like to log in again?
50X TAE (Tris-Acetate EDTA), pH 8.3
Supplier: GrowCells
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MRGF-4210-55GL MRGF-4210-020Q MRGF-4210-010Q MRGF-4211 MRGF-4210-010L MRGF-4210
75800-940EA 1000.25 USD InStock
75800-940 75800-942 75800-934 75800-944 75800-936 75800-938
50X TAE (Tris-Acetate EDTA), pH 8.3
Buffers Electrophoresis Buffers Running Buffers
TAE buffer is used in agarose gel electrophoresis and Polyacrylamide gels both as a buffer and a component of a gel
Can be used for both genomic and large supercoiled DNA. TAE buffer has a relatively low buffering capacity therefore buffer should be replaced periodicly during long electrophoresis runs. Offered in different concentration for all common biological applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty