Would you like to log in again?
10X TBE (Tris-Borate EDTA) pH 8.3
Supplier: GrowCells
MRGF-4216 MRGF4216010L MRGF4216020Q MRGF-4216-55GL MRGF-4215 MRGF4216010Q
75800-950EA 224.04 USD InStock
75800-950 75800-952 75800-954 75800-956 75800-946 75800-948
10X TBE (Tris-Borate EDTA) pH 8.3
Buffers Electrophoresis Buffers Running Buffers
TBE is the most commonly used buffer for DNA and RNA electrophoresis
Since borate is a strong inhibitor of enzymes, TBE is recommended for DNA/RNA sizing, not for recovery of DNA for enzymatic applications. Routinely used for DNA automated sequencing gels. Used both as a running buffer and gel preparation buffer. Recommended for use with fragments smaller than 1500 bp.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty