Would you like to log in again?
20X PBS (Phosphate Buffered Saline), pH 7.4
Supplier: GrowCells
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MRGF-6395 MRGF-6395-010L MRGF-6395-55GL MRGF-6396 MRGF-6395-020Q
75801-010EA 395.27 USD InStock
75801-010 75801-008 75801-006 75801-016 75801-014
20X PBS (Phosphate Buffered Saline), pH 7.4
Buffers Cell Culture Buffers
Balanced salt solution used in many biomolecular applications
Ultra pure quality makes buffer ideal for use in a variety of cell culture procedures, such as washing and transporting cells or tissue samples, diluting cell for counting and general preparation of reagents. PBS contains no Ca or Mg ions to provide efficient washing of chelators. Sold in 20X concentration to suit a variety of working dilution for any biological need.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty