Would you like to log in again?
20X SSC Buffer (Standard Sodium Citrate)
Supplier: GrowCells
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
MRGF-6495 MRGF-6495-020Q MRGF-6496 MRGF-6495-010Q MRGF-6495-010L MRGF-6495-200L
75801-022EA 316.76 USD InStock
75801-022 75801-020 75801-018 75801-028 75801-026 75801-024
20X SSC Buffer (Standard Sodium Citrate)
Buffers Electrophoresis Buffers Hybridization Buffers
Concentrated sodium chloride-sodium citrate buffer functions by providing high ionic strength to optimize the rate of probe annealing to target nucleic acid
Working solutions can easily be prepared from concentrated 20X solution with DI water.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty