Would you like to log in again?
Detonox™ Ultimate Precision Cleaner
Supplier: Alconox
Detonox™
2301 2355 2305
75801-040CS 463.51 USD InStock
75801-040 10015-626 10015-628
Detonox™ Ultimate Precision Cleaner
Cleaning Agents Detergents and Surface Cleansers
Detonox is a non-caustic detergent for exceptional removal of botanical, cosmetic, and pharmaceutical polymers, sticky extracts, resins, creams and lotions. Concentrated, highly emulsifying and penetrating formula is free rinsing, and leaves no residues.
  • Concentrated
  • Exceptional cosmetic and pharmaceutical polymer, cream and lotion removal
  • Non-caustic for greater worker safety and greater material compatibility
  • Free rinsing to give you reliable results and no interfering residues
Can be used to remove cosmetic bases and formulations, organics, grease, tars, sticky extracts, resins, waxes, fats, oils, titanium and zinc oxides, ointments and lotions.
Recommended for glass, stainless steel, PTFE, ceramic, cement and solvent resistant plastics and metals. Corrosion testing is advisable.
Cleaning method: Soak, circulate under immersions, ultrasonic cleaners, and manual cleaning.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty