Would you like to log in again?
COLUMN HPLC HPS C18 5UM 21.2X75MM
Supplier: MicroSolv Technology Corp
HPLC Column 5um 21.2mm x 75mm^ HPLC Column, Phase: Cogent HPS C18 High Purity Silica, Pore Size: 120A, Particle Size: 5um, Includes: Monofunctional bonding, An incredibly uniform surface area and structure, always perfectly spherical, for separation of compounds, Dimensions: 21.2mm x 75mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN HPLC HPS C18 5UM 21.2X75MM 75801-122 75018-P75 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty