Would you like to log in again?
COLUMN SEMI-PREP PHENYL COGENT 10X250 MM
Supplier: MicroSolv Technology Corp
HPLC Column 5um x 10mm^ HPLC Column, Phase: Cogent HPS Phenyl, Semi-Preparative column, Pore size: 120A, Particle size: 5um, High purity Silica, High efficiency, Application: For all types of analytes including bases, neutrals and acids, Inner dimension: 10mm, Length: 250mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN SEMI-PREP PHENYL COGENT 10X250 MM 75801-320 75020-SP250 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty