Would you like to log in again?
INSERT RUBBER FOR TUBES 10 ML
Supplier: HETTICH INSTRUMENTS
Rubber insert 13 x 65mm/15 x 75mm^ Rubber insert, Tube capacity: 2.6-5.5ml, Tube dimension: 13 x 65mm/15 x 75mm, Maximum RCF: 2,683, For swing out rotor, Rotina 380/380 RBenchtop Centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INSERT RUBBER FOR TUBES 10 ML 75801-926 0716 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty