Would you like to log in again?
ADAPTER F/1 PLACE ROTOR 93 X 134MM TUBE
Supplier: HETTICH INSTRUMENTS
Adapter 1-place^ Adapter, for Rotanta 460 Swing out rotor, 1-place, Maximum RCF: 4,779, Tube capacity: 600 ml, Dimension: 93x134 mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/1 PLACE ROTOR 93 X 134MM TUBE 75801-964 4846 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty