Would you like to log in again?
CARD FILTER 12.4 MM 1-4 ML SLIDE CARRIER
Supplier: HETTICH INSTRUMENTS
Filter card 6.2 / 30mm Capacity 1-4 ml Dimension: 12.4mm for slide carrier with ring^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CARD FILTER 12.4 MM 1-4 ML SLIDE CARRIER 75802-087 1675 Pack of 200 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty