Would you like to log in again?
FILTER CARD F/3X2ML 8.7/60MM CYTOCHAMBER
Supplier: HETTICH INSTRUMENTS
Filter card, Cyto chamber Capacity: 3x2ml, Dimension: 3x8.7mm, for slide carrier with ring^
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER CARD F/3X2ML 8.7/60MM CYTOCHAMBER 75802-090 1677 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty