Would you like to log in again?
ADAPTER F/1.1 -5ML BLOOD TUB 13.5X60.7MM
Supplier: HETTICH INSTRUMENTS
Adapter 13 x 90mm^ Adapter, tube capacity: 4.9ml, Tube Dimensions: 13 x 90mm, Maximum RCF: 2952, for Rotina 420/420 R Benchtop centrifuge, Swing-out rotor
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/1.1 -5ML BLOOD TUB 13.5X60.7MM 75802-246 5137 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty