Would you like to log in again?
ADAPTER F/4-7ML 16X75 TUBES SWG OUT RTR
Supplier: HETTICH INSTRUMENTS
Adapter 16 x 75mm^ Adapter, tube capacity: 4-7ml, Tube Dimensions: 16 x 75mm, Maximum RCF: 3005, for Rotina 420/420 R Benchtop centrifuge, Swing-out rotor
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/4-7ML 16X75 TUBES SWG OUT RTR 75802-248 5121-93 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty