Would you like to log in again?
ADAPTER FOR ROTOR 15 ML TUBES SET OF 6
Supplier: HETTICH INSTRUMENTS
Adapter^ Adapter, blood collection/urine tubes, tube capacity: 15ml, Dimension: 17x120mm, maximum RCF: 15,455, for Angle rotor, 12-place, UNIVERSAL 320
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER FOR ROTOR 15 ML TUBES SET OF 6 75802-310 1647 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty