Would you like to log in again?
ADAPTER F/ TWO TIER CYTO SLIDE SWING OUT
Supplier: HETTICH INSTRUMENTS
Two-tier frame^ Two-tier frame for cyto accessories, in large benchtop centrifuges or floor-standing centrifuges: Up to 16 cytological preparations can be obtained in one run, For Cyto chambers
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/ TWO TIER CYTO SLIDE SWING OUT 75802-324 5280 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty