Would you like to log in again?
ADAPTER F/25/30ML TUB ROUND/SKIRT 7 EACH
Supplier: HETTICH INSTRUMENTS
Adapter 25/30ml^ Adapter, Tube capacity: 25/30ml, Tube Dimensions: 25 x 90mm/25 x 110mm, Maximum RCF: 4,585, for Swing-out rotor, ROTINA 420 centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/25/30ML TUB ROUND/SKIRT 7 EACH 75802-394 4764 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty