Would you like to log in again?
ADAPTER 2-PLACE FOR 100ML TUBES 44X100MM
Supplier: HETTICH INSTRUMENTS
Adapter 44 x 100 mm^ Adapter, tube capacity: 100ml, Tube Dimensions: 44 x 100 mm, max. RCF 4,637, for Swing-out rotor, 2- places, ROTANTA 460/460 R centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER 2-PLACE FOR 100ML TUBES 44X100MM 75802-474 4737 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty