Would you like to log in again?
BUCKET RECTANGULAR TUBE F/BENCHTOP ROTOR
Supplier: HETTICH INSTRUMENTS
Bucket Swing-out rotor for ROTINA 460 centrifuge^ Bucket Swing-out rotor, without carriers, max. RCF 4,211, run-up in 90 second, run-down in 95 second, braked, for ROTINA 460 centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BUCKET RECTANGULAR TUBE F/BENCHTOP ROTOR 75802-510 5628 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty