Would you like to log in again?
ADAPTER FOR 50ML TUBES SKIRTED CONICAL
Supplier: HETTICH INSTRUMENTS
Adapter 29 x 115mm^ Adapter, conical, Tube capacity: 50ml, Tube Dimensions: 29 x 115mm, Maximum RCF: 4,187, for Swing-out rotor
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER FOR 50ML TUBES SKIRTED CONICAL 75802-520 6339-A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty