Would you like to log in again?
ADAPTER F/4-10ML 13X100MM HOLDS 47 TUBES
Supplier: HETTICH INSTRUMENTS
Adapter 13 x100mm^ Adapter, holds 47 tubes, tube capacity: 4-10ml, dimension: 13 x100mm, max. RC 2,906, for Swing-out rotor, ROTANTA 460/460 R centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/4-10ML 13X100MM HOLDS 47 TUBES 75802-538 5685 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty