Would you like to log in again?
ADAPTER F/175-290ML TUBE HOLDS 2 BOTTLES
Supplier: HETTICH INSTRUMENTS
Adapter holds 2 bottles^ Adapter, tube capacity: 175-290ml, Falcon*, Nalgene*, Nunc, with various dimension:61x118mm, 62 x144mm, 60 x130mm, 61x137mm, 61x122mm, 62 x137mm, max. RC 3,196, for Swing-out rotor, ROTANTA 460/460 R centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER F/175-290ML TUBE HOLDS 2 BOTTLES 75802-544 5681 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty