Would you like to log in again?
INSERT BALANCING F/CENT BLOOD BAG SINGLE
Supplier: HETTICH INSTRUMENTS
Balancing insert for blood bag^ Balancing insert for maximum of 750 ml blood bag system (4592-B), used in Swing-out rotor, if there are not enough blood bag systems to occupy every carrier of the rotor, empty carriers can be filled with balancing insert, Taring weight are used for balancing
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
INSERT BALANCING F/CENT BLOOD BAG SINGLE 75802-670 4587-A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty