Would you like to log in again?
BUCKET POLYPROPYLENE 96 X 135 MM 750 ML
Supplier: HETTICH INSTRUMENTS
Bucket 750 ml 96 x 135 mm^ Bucket, Capacity: 750 ml, Dimension: 96 x 135 mm, for Swing-out rotor, used in ROTO SILENTA 630 RS and ROTIXA 500 RS Floorstanding and ROTANTA 460/460 R Benchtop Centrifuge
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BUCKET POLYPROPYLENE 96 X 135 MM 750 ML 75802-682 4234-A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty