Would you like to log in again?
ADAPTER 1-PLACE FOR SWG OUT ROTOR 4-TIME
Supplier: HETTICH INSTRUMENTS
Adapter for blood bag triplicate^ Adapter for blood bags, triplicate, 1 place, maximum RCF: 5,252, run-up in second: 115, run-down in second braked 116, for Swing-out rotor, 4-place, used in ROTIXA 500RS Floorstanding centrifuge, Capacity: 450-500ml
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER 1-PLACE FOR SWG OUT ROTOR 4-TIME 75802-686 4244-A Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty