Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hettich EBA 270 Clinical Tabletop Centrifuges, Hettich Lab Technology

Supplier: Hettich

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

2300-01 270COMPLETE
75802-808EA 2816.92 USD InStock
75802-808 10812-622
Hettich EBA 270 Clinical Tabletop Centrifuges, Hettich Lab Technology
Centrifuges Benchtop Centrifuges
The Hettich Lab Technology EBA 270 clinical tabletop centrifuge features a built-in 90° swing-out rotor for quick and easy flat separation.

  • Max capacity- 6 x 15 mL
  • Max RPM/RCF – 4,000/2,254
  • Blood, urine, and GEL TUBES
  • Ideal for physicians and veterinarians
  • Built-in swing-out rotor for flat separation
  • Quiet operation – quiet as a normal conversation
  • Will not increase noise levels or contribute to distractions
  • Functional design – easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety – self-closing, auto-locking lid with steel latches
  • All-metal chamber provides reinforced security
  • Five year warranty – all Hettich centrifuges are protected by our 5-year warranty and full technical support
  • Fully certified – engineered and manufactured in accordance with all relevant safety and production standards
  • Made in Germany – German engineered and manufactured quiet, reliable and safe since 1904

Supports blood and round bottom tubes up to 15 mL and GEL TUBES

Complete application packages are available. Includes everything you need to perform your application: centrifuge, rotor, and adapters.

Hettich Lab Technology is an industry-leading laboratory equipment manufacturer based in Tuttlingen, Germany. Since 1904 we have designed, engineered, and manufactured precision equipment for the modern laboratory. Known for our vast array of centrifuges and incubators, Hettich delivers on quality, safety, and reliability.

Ideal for physicians' and veterinary offices.

Certifications: Hettich Lab Technology centrifuges are manufactured under all relevant national and international safety standards and certifications.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty