Would you like to log in again?
ADAPTER 1-PLACE W/ROTOR BUCKET F/13X75MM
Supplier: HETTICH INSTRUMENTS
Adapter for blood collection/urine tube^ Adapter for blood collection/urine tube, 1 place, Capacity: 1.6-5 ml, Dimension: 13x75 mm, RPM/RCF: less than or equal to 4,669/5,071, for 90 degree swing-out rotor, 6-place used in EBA 280 Tabletop Clinical Centrifuge, consistent platelet counts in minute
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTER 1-PLACE W/ROTOR BUCKET F/13X75MM 75802-828 1053 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty