Would you like to log in again?
VWR® B2-Series Analytical and Precision Balances
Supplier: VWR
VWR®
VWR-210B2 VWR-203B2 VWR-5002B2 VWR-120B2 VWR-64B2 VWR-124B2 VWR-403B2 VWR-60B2 VWR-503B2 VWR-214B2 VWR-4002B2
75802-868EA 1841.11 USD InStock
75802-868 75802-858 75802-864 76519-306 75802-866 76519-304 75802-860 75802-870 75802-862 76519-308 75802-872
VWR® B2-Series Analytical and Precision Balances
Balances and Scales
VWR® B2-Series balances combine advanced weighing features with intuitive operation.
  • Reliable results
  • External calibration
  • Multiple weigh modes
  • Worry-free 5-year warranty
  • 3-door draft shield included on analytical and precision models
VWR® B2-Series balances incorporate state-of-the-art features for advanced weighing applications. These economical balances offer a variety of standard features, such as external calibration, menu-driven operation, multiple weigh modes, and various application modes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty