Would you like to log in again?
CYTO-ID® Autophagy Detection Kit 2.0, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT175-0200 KIT175-0050
75802-912EA 311 USD InStock
75802-912 75802-914
CYTO-ID® Autophagy Detection Kit 2.0, Enzo Life Sciences
Assays Cellular Assays
A brighter and more photostable, no-transfection, quantitative assay for monitoring autophagy in live  cells.
  • No transfection assay eliminates need for transfection efficiency validation
  • Brighter, more photostable dye specifically stains autophagic vesicles
  • Negligible staining of lysosomes reduces background seen with other dyes
  • Rapidly quantifies autophagy in native heterogeneous cell populations
  • Facilitates high-throughput screening of activators and inhibitors of autophagy
The CYTO-ID® Autophagy detection kit 2.0 provides a rapid, specific, and quantitative approach for monitoring autophagy in live cells by fluorescence microscopy, flow cytometry, and microplate reader.
CYTO-ID® Autophagy Detection Kit 2.0 measures autophagic vacuoles and monitors autophagic flux in lysosomally inhibited live cells using a novel dye that selectively labels accumulated autophagic vacuoles. The dye has been optimized through the identification of titratable functional moieties that allow for minimal staining of lysosomes while exhibiting bright fluorescence upon incorporation into pre-autophagosomes, autophagosomes, and autolysosomes (autophagolysosomes). The assay offers a rapid and quantitative approach to monitoring autophagy in live cells without the need for cell transfection.Mechanism of ActionThe probe is a cationic amphiphilic tracer (CAT) dye that rapidly partitions into cells in a similar manner as drugs that induce phospholipidosis. Careful selection of titratable functional moieties on the dye prevents its accumulation within lysosomes, but enables labeling of vacuoles associated with the autophagy pathway.
Ordering information: Contains CYTO-ID® Green Detection Reagent 2Hoechst 33342 Nuclear StainAutophagy Inducer (Rapamycin)Chloroquine Control10X Assay Buffer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty