Would you like to log in again?
LDH Cytotoxicity WST Assay, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT157-2500 KIT157-0500
75802-922EA 286 USD InStock
75802-922 75802-924
LDH Cytotoxicity WST Assay, Enzo Life Sciences
Assays Cellular Assays
Colorimetric kit for the determination of cytotoxicity through the measurement of lactate dehydrogenase activity.
  • Homogenous assay using living cells or non-homogenous assay protocol using cell culture supernatant
  • Stable working solution
The LDH cytotoxicity WST assay is a colorimetric assay kit used to determine cytotoxicity by measuring lactate dehydrogenase activity released from damaged cells.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty