Would you like to log in again?
BNP Fragment ELISA Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
KIT152-0001
75802-928EA 736 USD InStock
75802-928
BNP Fragment ELISA Kit, Enzo Life Sciences
Assays ELISAs
Sensitive BNP fragment ELISA kit enabling detection of BNP fragment in serum and plasma.
  • Quantitative determination of human BNP fragment
  • Colorimetric, competitive enzyme immunoassay
For the quantitative determination of human BNP fragment in serum and plasma.
The BNP Fragment ELISA kit is a colorimetric competitive enzyme immunoassay kit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty