Would you like to log in again?
CRITERION™ Barney Miller Broth with Indicator, Hardy Diagnostics
Supplier: Hardy Diagnostics
C8991
75802-966EA 268.63 USD InStock
75802-966
CRITERION™ Barney Miller Broth with Indicator, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Barney Miller Broth with Indicator for the isolation and detection of beer spoilage microorganisms
  • Wide mouth opening - reduces dust formation by allowing the easy entry of a scoop
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers - for efficient storage
  • Increased solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Competitive pricing - call for a price quote for large volume purchases!
  • Complete technical/instruction sheets - available in HUGO software
  • Unparalleled performance - tested for optimal growth
Hardy Diagnostics CRITERION™ Barney Miller Broth with Indicator is recommended for the isolation and detection of beer spoilage microorganisms, specifically lactic acid bacteria
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards.
Container sizes include: - 2L mylar® bag – pre-measured for a 2 liter batch, no weighing needed!
- 500gm bottle - wide mouth, easy grip bottle, comes with a free scoop!
- 2kg and 10kg bucket - with convenient screw lid
Also see our selection of petri plates, glass screw-cap tubes, animal blood, media supplements and antibiotics.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty