Would you like to log in again?
HardyDisk™ AST Ceftazidime/Avibactam (CZA50), Avycaz (30 µg/20 µg), Hardy Diagnostics
Supplier: Hardy Diagnostics
Z9355 Z9351
75803-002EA 36.16 USD InStock
75803-002 75803-004
HardyDisk™ AST Ceftazidime/Avibactam (CZA50), Avycaz (30 µg/20 µg), Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST)
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST) also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Hardy Diagnostics offers a complete line of all commonly used antibiotics.
Available in single cartridges(Z9351) or packs of five. (Z9355) Compatible with the BBL dispensers.
Improved cartridge design helps to prevent jamming and tearing.
HardyDisk™ AST cartridges are packaged in an all new canister that features a desiccant built into the lid.
Kirby-Bauer accessories available; Mueller Hinton Agar, McFarland Standard, calipers, and much more!
Spring loaded in cartridges of 50 disks for smooth and reliable dispensing.
The last disk in each cartridge is clearly marked with dark lines around its edge to signify the need for refill.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty