Would you like to log in again?
ALP Urea, Modified, Hardy Diagnostics
Supplier: Hardy Diagnostics
Y70
75803-052PK 191.53 USD InStock
75803-052
ALP Urea, Modified, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
ALP Urea, Modified
(Aerobic Low Peptone) For the identification of non-fermentative bacilli. See technical insert.
Ordering information: Minimum Order - 5 packs
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty