Would you like to log in again?
SIEVE RING 360 DEG TRPZ HOL SS 1.0 MM
Supplier: VERDER SCIENTIFIC INC.
Grinding sieve for the SR 300^ Grinding sieve for the SR 300. Sieve has a trapezoidal design for efficient communition of material, 1.00mm
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SIEVE RING 360 DEG TRPZ HOL SS 1.0 MM 75803-164 024070032 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty