Would you like to log in again?
M17 Broth Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
GM1029-500G M1029-500G M1029-500G GM1029-5KG GM1029-2.5KG
75803-322EA 311.47 USD InStock
75803-322 75803-320 CA95020-326 95020-326 75803-324
M17 Broth Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and isolation of lactic streptococci and bacteriophages.
  • Microbiological culture media
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty