Would you like to log in again?
Lowenstein Jensen Medium Base, Granulated (L.J. Medium, Granulated), HiMedia Laboratories
Supplier: HiMedia
GM162-5KG GM162-2.5KG GM162-500G
75803-394EA 210.39 USD InStock
75803-394 75803-392 75803-396
Lowenstein Jensen Medium Base, Granulated (L.J. Medium, Granulated), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
This medium supports the growth of a wide variety of Mycobacteria and can also be used for niacin testing. This medium can be employed to identify saprophytes at room temperature (25°C) and also to identify presence or absence of pigmentation by photochromogenes and scotochromogenes at 35°C alternately in light and dark as per the type of organism.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty