Would you like to log in again?
Hektoen Enteric Agar, HiMedia
Supplier: HiMedia Laboratories
GM467-5KG GM467-500G GM467-2.5KG
75803-542EA 936.72 USD InStock
75803-542 75803-546 75803-544
Hektoen Enteric Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For differential and selective isolation of Salmonella and Shigella species from enteric pathological specimens.
  • Currently recommended as one of several plating media for the culture of Enterobacteriaceae from stool specimens
  • Multi-step procedure to isolate Salmonella
Hektoen Enteric Agar is currently recommended as one of several plating media for the culture of Enterobacteriaceae from stool specimens. Foods containing poultry, eggs or dairy
products are the most frequent vehicles for foodborne Salmonellosis, and a variety of procedures have been developed using Hektoen Enteric Agar as part of the multi-step procedure to isolate Salmonella.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty