Would you like to log in again?
Lauryl Sulphate Broth, HiMedia
Supplier: HiMedia Laboratories
GM080-500G GM080-2.5KG GM080-5KG
75803-702EA 980.94 USD InStock
75803-702 75803-700 75803-698
Lauryl Sulphate Broth, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Lauryl sulphate broth is a granulated media used for detection and enumeration of coliform bacteria in water, wastewater, dairy products and other food samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty