Would you like to log in again?
Dehydrated Media, Soyabean Casein Digest Medium (Tryptone Soya Broth), HiMedia Laboratories
Supplier: HiMedia Laboratories
GM011-5KG GM011-2.5KG GM011-500G
75803-726EA 732.01 USD InStock
75803-726 75803-724 75803-722
Dehydrated Media, Soyabean Casein Digest Medium (Tryptone Soya Broth), HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Soyabean casein digest medium, granulated is a general purpose medium used for cultivation of a wide variety of microorganisms and recommended for sterility testing of moulds and lower bacteria.
  • Cream to yellow coloured granular medium
  • pH of 3.0% weight/volume aqueous solution at 25 °C
Store below 30 °C in a tightly closed container and the prepared medium at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty