Would you like to log in again?
Diluent for Maximum Recovery, HiMedia Laboratories
Supplier: HiMedia Laboratories
GM1030-2.5KG GM1030-500G GM1030-5KG
75803-924EA 1330.91 USD InStock
75803-924 75803-922 75803-920
Diluent for Maximum Recovery, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
It is used for a protective and isotonic diluent used for maximal recovery of microorganisms.
  • Recommended for use as an isotonic dilutent
  • Combines protective effect of peptic digest of animal tissue with the osmotic balance of physiological saline
  • Maintain organisms for one to two hours of dilution without multiplication
Maximum Recovery Diluent is formulated as recommended by ISO Committee for use as an isotonic diluent
Maximum Recovery Diluent combines protective effect of peptic digest of animal tissue with the osmotic balance of physiological saline. The low concentration of peptic digest of animal tissue helps to maintain the organisms for one to two hours of dilution without multiplication. The isotonic property of the diluent ensures the recovery of organisms from various sources, which may be vulnerable in distilled water or aqueous suspensions.
Put 10 gm of test sample into a sterile blender jar and add 90 ml of sterile Maximum Recovery Diluent. Operate the blender at 15,000 to 20,000 revolutions per minute. Transfer 1mL of it to 9mL of sterile diluent within 15 minutes and mix well. It will be 10 to 1 dilution. Appropriate serial dilutions can be prepared using same diluent and counts obtained by spread plate or pour plate technique. Use a positive test sample divided between new and previous diluent. Carry out duplicate tests as described in technique and look for equivalent yields of organisms between the diluent batches.
Incubate the tubes with test organisms. At time zero and after 30 minutes at room temperature, subculture a loopful (0.01 ml) onto Soyabean Casein Digest Agar (M290) with 5% v/v sheep blood using streak technique. Incubate plates at 35 ± 2°C for 18 to 24 hours.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty