Would you like to log in again?
Tartoff - Hobbs (Terrific) Broth, HiMedia Laboratories
Supplier: HiMedia Laboratories
GM1250-500G GM1250-5KG GM1250-2.5KG
75804-052EA 799.99 USD InStock
75804-052 75804-054 75804-056
Tartoff - Hobbs (Terrific) Broth, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Tartoff-Hobbs (Terrific) broth is used for the cultivation of recombinant strains of Escherichia coli.
  • Cream to light yellow coloured
  • Granular medium
  • Reaction of 4.76% w/v aqueous solution at 25 °C
Caution: Store below 30 °C in tightly closed container and the prepared medium at 2 to 8 °C.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty