Would you like to log in again?
King’s Medium B Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M1544-500G M1544-500G GM1544-2.5KG GM1544-5KG GM1544-500G
95021-270EA 213.7 USD InStock
95021-270 CA95021-270 75804-172 75804-174 75804-176
King’s Medium B Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the nonselective isolation, cultivation, and pigment production of Pseudomonas species.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty